missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC15A4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5107259

Product Code. 17157063

  • 586.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

SLC15A4 is the proton oligopeptide cotransporter. It transports free histidine and certain di- and tripeptides.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC15A4
Polyclonal
Unconjugated
SLC15A4
AA987064; AW742963; C130069N12Rik; FP12591; hPHT1; Peptide transporter 4; peptide/histidine transporter 1; peptide-histidine transporter 4; Pht1; PTR4; rPHT1; Slc15a4; solute carrier family 15 (oligopeptide transporter), member 4; solute carrier family 15 member 4; solute carrier family 15, member 4
Rabbit
Affinity Chromatography
RUO
121260
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q8N697
SLC15A4
Synthetic peptide directed towards the middle region of human SLC15A4 (aa 396-446).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.