Learn More
SLC17A9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26731-1-AP-150UL
 View more versions of this product
Product Code. 16895105
  • 410.00€ / 150µL
Description
Involved in vesicular storage and exocytosis of ATP. May accumulate ATP and other nucleotides in secretory vesicles such as adrenal chromaffin granules and synaptic vesicles.
Specifications
Specifications
SLC17A9
Polyclonal
Unconjugated
SLC17A9
C20orf59, SLC17A9, Vesicular nucleotide transporter, VNUT
Rabbit
Antigen Affinity Chromatography
RUO
228993, 63910
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VCL5, Q9BYT1
SLC17A9
SLC17A9 Fusion Protein Ag24970
150 μL