missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC1A5 Antibody (idactamab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28813-1mg
This item is not returnable.
View return policy
Description
SLC1A5 Monoclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SLC1A5 | |
| Monoclonal | |
| Unconjugated | |
| Q15758 | |
| Human | |
| Protein A purified | |
| RUO | |
| 6510 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| idactamab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
| SLC1A5/ ASCT2 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido