missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | SLC1A5 |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603512
|
Novus Biologicals
NBP2-97567-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661445
|
Novus Biologicals
NBP2-97567-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC1A5 Polyclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC1A5 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
| Produced in rabbits immunized with E. coli-derived Human SLC1A5 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 6510 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title