missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC20A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144916
This item is not returnable.
View return policy
Description
Immunogen sequence: YTSYCNAVSD LHSASEIDMS VKAEMGLGDR KGSNGSLEEW YDQDKPEVS Antigen sequence identity: Mouse 94%; Rat 94%.
Retrovirus receptors allow infection of human and murine cells by various retroviruses. The receptors that have been identified at the molecular level include CD4 for human immunodeficiency virus, Rec1 for murine ecotropic virus, and GLVR1 for gibbon ape leukemia virus. These 3 proteins show no homology to one another at the DNA or protein level. GLVR1 is a sodium-dependent phosphate symporter.
Specifications
| SLC20A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC20A1 | |
| AI607883; DKFZp686J2397; FLJ41426; Gibbon ape leukemia virus receptor 1; GLVR1; Glvr-1; leukemia virus receptor 1 homolog; Phosphate transporter 1; PIT1; piT-1; RPHO-1; Slc20a1; Sodium-dependent phosphate transporter 1; Sodium-dependent phosphate transporter 1-like protein; solute carrier family 20 (phosphate transporter), member 1; solute carrier family 20 member 1; solute carrier family 20, member 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6574 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUM9 | |
| SLC20A1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 464-512. Recombinant protein control fragment (Product #RP-110199). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction