Learn More
SLC22A3 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111769
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)ERRGQGH (S)HRYLLE Homology: Rat, mouse - 13/14 amino acid residues identical.
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq, Jul 2008].
Specifications
| SLC22A3 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| SLC22A3 | |
| 3-Oct; EMT; EMTH; Extraneuronal monoamine transporter; OCT3; Orct3; Organic cation transporter 3; Slc22a3; Slca22a3; Solute carrier family 22 member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20519, 29504 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O88446, Q9WTW5 | |
| SLC22A3 | |
| Peptide corresponding to amino acid residues 83-96 of rat OCT3 (Extracellular, 1 st loop) | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.