missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated pan Ras Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60309-20UL
This item is not returnable.
View return policy
Description
The pan Ras antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human pan Ras. This antibody recognizes human, mouse, rat, pig antigen. The pan Ras antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| pan Ras | |
| Monoclonal | |
| 1000 μg/mL | |
| C-K-RAS; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS1; KRAS2; NS3; RASK2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Conjugated Antibody | |
| IgG1 |
| Flow Cytometry (Intracellular Staining) | |
| 4H4G7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3845 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction