missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A6 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98638FR
331.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
SLC26A6 Polyclonal antibody specifically detects SLC26A6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
SLC26A6 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
DyLight 680 | |
anion transporter 1, DKFZp586E1422, pendrin L1, pendrin-L1, Pendrin-like protein 1, solute carrier family 26 member 6, solute carrier family 26, member 6, sulfate anion transporter | |
Produced in rabbits immunized with E. coli-derived Human SLC26A6 fragment. | |
0.1 mL | |
Endocrinology, Signal Transduction | |
65010 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction