Learn More
PD-L1 Antibody (Hu125) [Allophycocyanin], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals FAB10355A
Product Code. 18646713
  • 664.00€ / 0.10mL
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-L1
Monoclonal
APC
PBS
Human
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
Hu125
Flow Cytometry, CyTOF-ready
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Human PD-L1/B7-H1,
0.1 mL
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
29126
Store at 4°C in the dark.