Learn More
SLC2A9 Mouse anti-Human, Unconjugated, Clone: 2D6E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67530-1-IG-150UL
 View more versions of this product
Product Code. 16810216
  • 410.00€ / 150µL
Description
The SLC2A9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC2A9. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SLC2A9 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SLC2A9
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry, Immunofluorescence
2D6E1
Unconjugated
150 μL
56606