missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76498AF647
This item is not returnable.
View return policy
Description
SLC39A1 Polyclonal antibody specifically detects SLC39A1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLC39A1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| CGI-08, CGI-71, IRT1, solute carrier family 39 (zinc transporter), member 1, solute carrier family 39 (zinc transporter), member 3, ZIP1, ZIP-1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human ZIP1. The immunogen is located within amino acids 40 - 90 of ZIP1. Amino Acid Squence: LRRPGANHEGSASRQK | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 27173 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction