missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A7/ZIP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 624.00€
Specifications
| Antigen | SLC39A7/ZIP7 |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18471332
|
Novus Biologicals
NBP2-13338-25ul |
25ul |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18004838
|
Novus Biologicals
NBP2-13338 |
0.1 mL |
624.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC39A7/ZIP7 Polyclonal specifically detects SLC39A7/ZIP7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SLC39A7/ZIP7 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7922 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: GHDDLHDDLQEDFHGHSHRHSHEDFHHGHSHAHGHGHTHESIWHGHTHDHDHGHSHEDLHHG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| D6S115E, D6S2244EReally interesting new gene 5 protein, H2-KE4, HKE4Solute carrier family 39 member 7, HLA class II region expressed gene KE4, KE4, Ke4 gene, mouse, human homolog of, RING5Histidine-rich membrane protein Ke4, solute carrier family 39 (zinc transporter), member 7, zinc transporter SLC39A7, ZIP7 | |
| SLC39A7 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title