missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC5A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542485
This item is not returnable.
View return policy
Description
Peptide sequence: DDNGIYNQKF PFKTLAMVTS FLTNICISYL AKYLFESGTL PPKLDVFDAV Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
Specifications
| SLC5A7 | |
| Polyclonal | |
| Unconjugated | |
| Slc5a7 | |
| CHT; CHT1; hCHT; Hemicholinium-3-sensitive choline transporter; high affinity choline transporter 1; high affinity choline transporter 1; LOW QUALITY PROTEIN: high affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; HMN7A; MGC126299; MGC126300; similar to solute carrier family 5 (choline transporter), member 7; SLC5A7; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; solute carrier family 5 member 7; solute carrier family 5, member 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 60482 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9GZV3 | |
| Slc5a7 | |
| Synthetic peptide directed towards the middle region of human SLC5A7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction