missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13315-1-AP-20UL
This item is not returnable.
View return policy
Description
The yeast heterotetrameric GINS complex is made up of Sld5, Psf1 (GINS1), Psf2 (GINS2), and Psf3 (GINS3). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.Specifications
| SLD5 | |
| Polyclonal | |
| Unconjugated | |
| GINS4 | |
| GINS complex subunit 4, GINS4, SLD5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109145, 84296 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99LZ3, Q9BRT9 | |
| GINS4 | |
| SLD5 Fusion Protein Ag4158 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction