Learn More
Zeiss™ Slider
Product Code. 11835093
Change view
Click to view available options
Type:
Slider
Slider with Fixed Phase Stop Ph 1/0.4
Description:
DIC slider EC EP
DIC slider EC EP Epi+
Slider w/ Fixed Phase Stop Ph 1/0.4
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Description unitSize
11835093 Slider DIC slider EC EP Each
11825093 Slider DIC slider EC EP Epi+ Each
12312043 Slider with Fixed Phase Stop Ph 1/0.4 Slider w/ Fixed Phase Stop Ph 1/0.4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11835093 Supplier Zeiss™ Supplier No. 4269689030000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
Use these sliders for a variety of applications
Slider 10x46 mm Ph/PlasDIC, H, Ph/PlasDIC
  • 2 positions for Ph or PlasDIC diaphragms
  • brightfield position with neutral
  • density filter 0.06
Slider 10x46 mm
  • With fixed phase stop Ph 1/0.4
DIC slider EC EP
  • 5x/0.13, 10x/0.2, 20x/0.4 Epi+
DIC slider EC EP
  • 50x/0.7 Epi+
Specifications
Type Slider
Description DIC slider EC EP
Quantity 1 Ea.