Learn More
Horst™ Slim Heating Tape with High Specific Heat Output
Product Code. 10659042
Change view
Click to view available options
Length (Metric):
1.0 m
2.0 m
3.0 m
10.0 m
Wattage:
50 W
100 W
150 W
480 W
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Wattage Unit Size
10659042 1.0 m 50 W Each
10458641 2.0 m 100 W Each
10468641 3.0 m 150 W Each
10513282 10.0 m 480 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10659042 Supplier Horst™ Supplier No. 25202
  • ONLINE EXCLUSIVE PRICE
    124.13€
    77.46€ / Pack of 1
    Save 46.67€
    37% Off
View all promotions
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Double-laid heating conductors in a slim, insulated tape.
  • Slim design.
  • Electrically insulated with PTFE.
  • Metallic protective braid of nickel-plated copper.
Specifications
Description Bending radius: 10 mm, Thickness: 4 mm
Length (Metric) 1.0 m
Voltage 230 V
Material PTFE, Nickel-plated Copper
Length (Metric) Power Cord 1 m
Wattage 50 W
Width (Metric) 4 mm
Temperature (Metric) 250°C (Nominal)