missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLP-76/LCP2 Antibody (SLP-76/03), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-537UV
This item is not returnable.
View return policy
Description
SLP-76/LCP2 Monoclonal specifically detects SLP-76/LCP2 in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLP-76/LCP2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LCP2 | |
| Bacterially expressed fusion protein representing amino acids 216-434 of human SLP76 with histidine tag | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 3937 | |
| Human, Mouse, Pig | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SLP-76/03 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The polyclonal antibody reacts with SLP76, a 76kDa cytosolic adaptor protein that is involved in signaling of various hematopoietic cells, such as T cells, mast cells or neutrophils; in B cells, however, it is replaced by SLP65. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction