Learn More
SMAD7 Mouse, anti-Human, Clone: 3D1C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68594-1-IG-150UL
 View more versions of this product
Product Code. 18060736
  • 410.00€ / 150µL
Description
The SMAD7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMAD7. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SMAD7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SMAD7
3D1C3
CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MADH7, MADH8, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7, SMAD7
150 μL
4092
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Rabbit, Pig
150 kDa