Learn More
SMAD7 Mouse, anti-Human, Clone: 3D1C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68594-1-IG-150UL
Description
The SMAD7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMAD7. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SMAD7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SMAD7 | |
| 3D1C3 | |
| CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MADH7, MADH8, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7, SMAD7 | |
| 150 μL | |
| 4092 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 150 kDa | |
| IgG2a |