missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16597-1-AP-20UL
This item is not returnable.
View return policy
Description
SMAP2 (Small ArfGAP2) is a Protein Coding gene. Among its related pathways are Endocytosis. Gene Ontology (GO) annotations related to this gene include GTPase activator activity. An important paralog of this gene is SMAP1.Specifications
| SMAP2 | |
| Polyclonal | |
| Unconjugated | |
| SMAP2 | |
| RP1 228H13.3, small ArfGAP2, SMAP1L, SMAP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298500, 64744, 69780 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TN29, Q8WU79 | |
| Smap2 | |
| SMAP2 Fusion Protein Ag9751 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction