missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14185-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined.Specifications
| SMC3 | |
| Polyclonal | |
| Unconjugated | |
| SMC3 | |
| BAM, Bamacan, BMH, CDLS3, CSPG6, HCAP, SMC 3, SMC protein 3, SMC3, SMC3L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13006, 29486, 9126 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97690, Q9CW03, Q9UQE7 | |
| SMC3 | |
| SMC3 Fusion Protein Ag5440 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction