Learn More
Lonza™ SmGM™- 2 Smooth Muscle Cell Growth Medium-2 SingleQuots™ Supplements and Growth Factors
Product Code. 11655500
Change view
Click to view available options
Quantity:
1 mL hFGF-B, 25 mL FBS, 0.5 mL Insulin, hEGF and GA-1000
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11655500 1 mL hFGF-B, 25 mL FBS, 0.5 mL Insulin, hEGF and GA-1000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11655500 Supplier Lonza™ Supplier No. CC4149
  • ONLINE EXCLUSIVE PRICE
    346.00€
    300.21€ / Pack of 1
    Save 45.79€
    13% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Supplements and growth factors kit for use with smooth muscle cell basal medium.
  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full SmGM™-2 Smooth Muscle Cell Growth Medium BulletKit™ (catalog number CC-3182), which consists of basal medium and supplements packaged together.
  • Developed for maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more.
  • Also used to support the maintenance of intestinal cells.
  • Includes the following:
    • 1 x Lilac Cap Vial with Insulin 0.50 mL
    • 1 x Gray Cap Vial with hFGF-B 1.00 mL
    • 1 x Red Cap Vial with GA-1000 0.50 mL
    • 1 x Bottle FBS 25.00 mL
    • 1 x Green Cap Vial with hEGF 0.50 mL
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Smooth Muscle Cells
Product Type Supplements and growth factors
Quantity 1 mL hFGF-B, 25 mL FBS, 0.5 mL Insulin, hEGF and GA-1000
Content And Storage Insulin, hFGF-B, GA-1000, FBS, hEGF