missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03326AF594
This item is not returnable.
View return policy
Description
SMN Polyclonal antibody specifically detects SMN in Human samples. It is validated for Western Blot
Specifications
| SMN | |
| Polyclonal | |
| Western Blot | |
| Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
| The immunogen recognized by this antibody maps to a region between residue 244 and 294 of human survival of motor neuron 1 using the numbering given in entry NP_000335.1 (GeneID 6606). | |
| 0.1 mL | |
| Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators | |
| 6606 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction