Learn More
SMS Monoclonal Antibody (OTI 5F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503112
Description
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).Specifications
| SMS | |
| Monoclonal | |
| 0.36 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52788 | |
| SMS | |
| Full length human recombit protein of human SMS produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI 5F9 | |
| Unconjugated | |
| SMS | |
| MRSR, SPMSY, SRS, SpS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6611 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.