Learn More
SNAP25 Monoclonal Antibody (OTI 1D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502838S
Description
SNAP25 Monoclonal antibody specifically detects SNAP25 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| SNAP25 | |
| Monoclonal | |
| 0.65 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H0R4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84064 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI 1D5 | |
| Unconjugated | |
| HDHD2 | |
| ALMS1; bA416N4.2; Bdr; blind drunk; CG17676; CG17884; CG40452; CMS18; dJ1068F16.2; Dmel CG40452; Dmel_CG40452; etID17571.20; etID69167.20; fj42b02; fj45g04; fj48c02; FLJ23079; GENA70; HGNC:11132; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; si:dkeyp-8f4.6; Snap; SNAP 25; snap25; SNAP-25; snap25.1; SNAP-25.1; snap25a; SNAP-25A; SNAP-25B; snp25; sp; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein 25-A; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25a; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25a; synaptosome associated protein 25kDa; synaptosome-associated protein 25.1; synaptosome-associated protein 25a; wu:fj42b02; wu:fj45g04; wu:fj48c02 | |
| Full length human recombinant protein of human HDHD2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.