Learn More
SNaPshot™ Multiplex Kit
Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.
Brand:  ABI Applied Biosystems 4323155
Additional Details : Weight : 0.59420kg
 View more versions of this product
Product Code. 15899756
  • 28640.00€ / Each
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.
Order Info
Each
Specifications
Specifications
Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer
SNaPshot™ Multiplex Kit
5000