Learn More
SNaPshot™ Multiplex Kit
Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.
6560.00€ - 28640.00€
Specifications
Accession Number Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer
Description SNaPshot™ Multiplex Kit
View More Specs
Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15889756
View Documents Promotion Details
ABI Applied Biosystems
4323154
1000
6560.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
15899756
View Documents Promotion Details
ABI Applied Biosystems
4323155
5000
28640.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.
Order Info
Each
Specifications
Specifications
Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer
SNaPshot™ Multiplex Kit