Learn More
Chemsplash Snood Cap Peak Disposable
GREENER_CHOICE
Product Code. 30058382
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30058382 Blue Pack of 1000
30058431 White Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30058382 Supplier Chemsplash Supplier No. 2528.300.999
  • ONLINE EXCLUSIVE PRICE
    208.00€
    186.99€ / Pack of 1000
    Save 21.01€
    10% Off
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Snood Caps are a polypropylene heavyweight material that is also available in cotton or nylon.
Snood Caps
  • Heavyweight Polypropylene
  • PFAS Free
Description: Snood Caps are made from a heavyweight polypropylene material, and they are also available in cotton or nylon.
Specifications
Product Type Snood Cap
Size One Size
Material Polypropylene
Color Blue
Closure Type Ties
Disposable Single-use
Latex Free Yes