Learn More
Chemsplash Snood Cap Peak Disposable
Product Code. 30058431
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30058431 White Pack of 1000
30058382 Blue Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30058431 Supplier Chemsplash Supplier No. 2528.400.999
  • ONLINE EXCLUSIVE PRICE
    221.52€
    186.99€ / Pack of 1000
    Save 34.53€
    15% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Snood Caps are a polypropylene heavyweight material that is also available in cotton or nylon.
Snood Caps
  • Heavyweight Polypropylene
  • PFAS Free
Description: Snood Caps are made from a heavyweight polypropylene material, and they are also available in cotton or nylon.
Specifications
Product Type Snood Cap
Size One Size
Material Polypropylene
Color White
Closure Type Ties
Disposable Single-use
Latex Free Yes