missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRP70 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37896
This item is not returnable.
View return policy
Description
SNRP70 Polyclonal specifically detects SNRP70 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SNRP70 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P08621 | |
| SNRNP70 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RDPIPYLPPLEKLPHEKHHNQPYCGIAPYIREFEDPRDAPPPTRAETREERMERKRREKIERRQQEVETELKMWDPHNDPNAQ | |
| 0.1 mL | |
| Primary | |
| Specificity of human SNRP70 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RNPU1ZU170K, RPU1U1AP, small nuclear ribonucleoprotein 70kDa (U1), Snp1, snRNP70, SNRP70U1 small nuclear ribonucleoprotein 70 kDa, U1 snRNP 70 kDa, U1-70Ksmall nuclear ribonucleoprotein 70kDa (RNP antigen), U1AP1, U1RNP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6625 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction