missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPB Monoclonal Antibody (Y12), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513446
Additional Details : Weight : 0.01000kg
Description
The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.Specifications
SNRPB | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P14678, P27048 | |
SNRPB | |
Protein G | |
RUO | |
100341600, 100624586, 171365, 20638, 6628 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot | |
Y12 | |
Biotin | |
SNRPB | |
AL024368; AU018828; B polypeptide of Sm protein; CCMS; COD; sm protein B; Sm protein B/B'; SM11; small nuclear ribonucleoprotein B; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; small nuclear ribonucleoprotein polypeptides B and B1; small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; sm-B; Sm-B/B'; SmB/B'; sm-B/Sm-B'; SmB/SmB'; SNRNP-B; Snrpb; SNRPB1 | |
Mouse | |
500 μL | |
Primary | |
Drosophila, Human, Mouse, Porcine, Rabbit, Rat | |
Antibody | |
IgG3 κ |