missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521747
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| SNX18 | |
| Polyclonal | |
| Unconjugated | |
| SNX18 | |
| SH3 and PX domain-containing protein 3B; SH3PX2; SH3PXD3B; Snag1; Snx18; sorting nexin 18; sorting nexin associated golgi protein 1; sorting nexin-18; Sorting nexin-associated Golgi protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112574 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96RF0 | |
| SNX18 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SNX18. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion