missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX4 Recombinant Rabbit Monoclonal Antibody (24GB2335)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557734
This item is not returnable.
View return policy
Description
SNX4 Recombinant Monoclonal Antibody for Western Blot, Flow
SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors.
Specifications
| SNX4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SNX4 | |
| 1810036H14Rik; AI037066; ATG24B; Snx4; sorting nexin 4; sorting nexin-4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 360725, 69150, 8723 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2335 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95219, Q91YJ2 | |
| SNX4 | |
| A synthesized peptide derived from human SNX4 (aa 321-400). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction