Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Product Code. 11982188 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric):
75 mm
100 mm
150 mm
Diameter (Metric):
10 mm
12 mm
16 mm
18 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
11982188 75 mm 10 mm Pack of 100
12961031 100 mm 12 mm Pack of 100
11912198 150 mm 16 mm Pack of 100
11922198 150 mm 18 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11982188
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
  • Soda Lime Glass
  • Rimless
  • Disposable 
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 75 mm
Material Soda Lime Glass
Quantity 100 Pack
Rimmed No
Diameter (Metric) 10 mm