missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-hexadecanesulfonate, 97%

Product Code. 11414297
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414297 5 g 5g
11424297 25 g 25g
11434297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414297 Supplier Thermo Scientific Alfa Aesar Supplier No. B20382.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15015-81-3 | C16H33NaO3S | 328.487 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15015-81-3
Molecular Formula C16H33NaO3S
Molecular Weight (g/mol) 328.487
MDL Number MFCD00007528
InChI Key PNGBYKXZVCIZRN-UHFFFAOYSA-M
Synonym sodium cetylsulfonate, sodium 1-hexadecanesulfonate, 1-hexadecanesulfonic acid sodium salt, 1-hexadecanesulfonic acid, sodium salt, sodium hexadecane-1-sulfonate, unii-sk4e885p15, sodium hexadecane-1-sulphonate, cetylsulfonic acid sodium salt, 1-hexadecanesulfonic acid, sodium salt 1:1, hexadecylsulfonic acid sodium salt
PubChem CID 23665769
IUPAC Name sodium;hexadecane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Linear Formula CH3(CH2)15SO3Na
Quantity 5 g
Beilstein 3920654
Formula Weight 328.49
Percent Purity 97%
Chemical Name or Material Sodium 1-hexadecanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.