Learn More
Sodium Acetate-13C,D3, TRC
Product Code. 30423321
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423321 100 mg 100mg
30410474 50 mg 50mg
30418444 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30423321 Supplier Toronto Research Chemicals Supplier No. TRCS960210100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102212-93-1
Molecular Formula C13CD3NaO2
Molecular Weight (g/mol) 86.04
Synonym Acetic-1-13C-2,2,2-d3 Acid Sodium Salt
IUPAC Name sodium;2,2,2-trideuterioacetate
SMILES [2H]C([2H])([13C](O[Na])=O)[2H]
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1D3,2+1;
Formula Weight 86.0253
Chemical Name or Material Sodium Acetate-13C,D3