missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Arsenite solution EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
500 mL
Unit Size:
1000mL
Specifications
Specifications
| Packaging | Bottle |
| CAS | 7784-46-5 |
| Molecular Formula | AsNaO2 |
| Concentration | 0.1 M |
| InChI Key | PTLRDCMBXHILCL-UHFFFAOYSA-M |
| SMILES | [O-][As]=O.[Na+] |
| IUPAC Name | sodium;oxoarsinite |
| Molecular Weight (g/mol) | 129.909 |
| PubChem CID | 443495 |
| Quantity | 500 mL |
| Show More |
Safety and Handling
Product Identifier
- Sodium arsenite 0.1M. Volumetric Solution European Pharmacopoeia Ref: 3005800
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P330-Rinse mouth.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction