Learn More
Sodium Azide, TRC
Product Code. 30006259
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006259 10 g 10g
30006202 5 g 5g
30003881 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006259 Supplier Toronto Research Chemicals Supplier No. TRCS61253010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26628-22-8
Molecular Formula N3 . Na
Molecular Weight (g/mol) 65.01
Synonym Sodium Azide, Agrizide, Hydrazoic Acid Sodium Salt, Micro-O-Protect, NSC 3072, Nemazyd, SEP 100
IUPAC Name sodium;azide
SMILES [Na+].[N-]=[N+]=[N-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/N3.Na/c1-3-2;/q-1;+1
Formula Weight 64.999
Chemical Name or Material Sodium Azide