missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide (95%), TRC

Product Code. 30425796
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Product Code. Quantity unitSize
30425796 5 g 5g
30413601 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 30425796

Brand: Toronto Research Chemicals TRCS6146535G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium bis(trimethylsilyl)amide
CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Quantity 5 g
Synonym Hexamethyldisilazane sodium salt
InChI Formula InChI=1S/C6H18NSi2.Na/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1
SMILES [Na+].C[Si](C)(C)[N-][Si](C)(C)C
IUPAC Name sodium;bis(trimethylsilyl)azanide
Molecular Weight (g/mol) 183.37
Formula Weight 183.0875
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.