missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium borohydride, 99%, (VenPureTM SF granules), granules
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
100g
10kg
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 360.0°C |
| Quantity | 2.5 kg |
| Assay Percent Range | 98.5% min. (Hydrogen evolution) |
| Linear Formula | NaBH4 |
| Merck Index | 15,8727 |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents |
| Formula Weight | 37.83 |
| Percent Purity | 99% |
| Packaging | Plastic Bottle |
| Chemical Name or Material | Sodium borohydride |
Safety and Handling
Product Identifier
- Sodium borohydride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H301-Toxic if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction