Learn More
Sodium chlorate, 99% min, Thermo Scientific™
35.60€ - 176.65€
Chemical Identifiers
| CAS | 7775-09-9 |
|---|---|
| Molecular Formula | ClNaO3 |
| Molecular Weight (g/mol) | 106.44 |
| MDL Number | MFCD00003479 |
| InChI Key | YZHUMGUJCQRKBT-UHFFFAOYSA-M |
| Synonym | sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal |
| PubChem CID | 516902 |
| ChEBI | CHEBI:65242 |
| IUPAC Name | sodium;chlorate |
| SMILES | [O-]Cl(=O)=O.[Na+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11377857
|
Thermo Scientific Alfa Aesar
014267.22 |
100g |
35.60€
100g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11367857
|
Thermo Scientific Alfa Aesar
014267.A1 |
1kg |
52.20€
1kg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11387857
|
Thermo Scientific Alfa Aesar
014267.D9 |
5 x 1kg |
196.00€ 176.65€ / 5kg Save 19.35€ 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| ClNaO3 | |
| MFCD00003479 | |
| sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
| CHEBI:65242 | |
| [O-]Cl(=O)=O.[Na+] |
Specifications
| 7775-09-9 | |
| ClNaO3 | |
| sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
| YZHUMGUJCQRKBT-UHFFFAOYSA-M | |
| sodium;chlorate | |
| 516902 | |
| 106.45 | |
| Colorless granules | |
| Sodium chlorate |
| 99% min. | |
| MFCD00003479 | |
| Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia. | |
| [O-]Cl(=O)=O.[Na+] | |
| 106.44 | |
| CHEBI:65242 | |
| Odorless | |
| 2.5 g/cm3 |
Safety and Handling
P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
H271-H302-H373
missing translation for 'dotInformation' : Hazard Class: 5.1, Packing Group: II
missing translation for 'einecsNumber' : 231-887-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.