missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, 0.100 Normal (N/10), Ricca Chemical

Product Code. 31750066
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750066 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750066 Supplier Ricca Chemical Company Supplier No. R7224100500A
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Boiling Point 100°C
Melting Point 0.0°C
CAS 7732-18-5, 7647-14-5, 7647-14-5, 7732-18-5
Color Colorless
CAS Min % 99.42%, 0.58%
Physical Form Liquid
Quantity 500 mL
CAS Max % 0.58%, 99.42%
Molecular Weight (g/mol) 58.44 g/mol
Grade Laboratory
Packaging 500 mL HDPE Bottle
Appearance Colorless Liquid
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.