Learn More
Sodium chloride, Optical Grade
Description
Sodium Chloride (NaCl) is widely used in biochemistry and molecular biology applications. It is considered an essential nutrient. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. NaCl is a component of phosphate buffered saline and SSC buffer. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1413°C |
| Melting Point | 801°C |
| Color | White |
| pH | 6 to 9 |
| Physical Form | Broken Crystals |
| Quantity | 250 g |
| Merck Index | 14,8599 |
| Solubility Information | Soluble in water at 20°C 358g/L. Very slightly soluble in alcohol,glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid. |
| Formula Weight | 58.44 |
| Grade | Optical |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.