missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride p. For Laboratory Use, Chem Lab

Product Code. 13218253
Change view
Click to view available options
Sodium Chloride p. For Laboratory Use, 5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13218253 Sodium Chloride p. For Laboratory Use, 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13218253 Supplier Chem-Lab Supplier No. CL00.1423.5000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Specifications

Boiling Point 1413°C
Melting Point 801°C
CAS 7647-14-5
Color Colorless
Physical Form Solid
Quantity 5 kg
Assay Percent Range > 99.7%
Molecular Formula ClNa
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Grade Pure
Packaging Polyethylene
Density 2.17 g/cm3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.