missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloroacetate, 98%

Product Code. 11359216
Change view
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Unit Size:
10000g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359216 10,000 g 10000g
11413980 250 g 250g
11489523 500 g 500g
11499523 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11359216 Supplier Thermo Scientific Alfa Aesar Supplier No. A12379.0C
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3926-62-3 | C2H2ClNaO2 | 116.476 g/mol

Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

Solubility
Soluble in water, ether, chloroform, benzene and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3926-62-3
Molecular Formula C2H2ClNaO2
Molecular Weight (g/mol) 116.476
MDL Number MFCD00002684
InChI Key FDRCDNZGSXJAFP-UHFFFAOYSA-M
Synonym sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech
PubChem CID 23665759
IUPAC Name sodium;2-chloroacetate
SMILES C(C(=O)[O-])Cl.[Na+]

Specifications

Melting Point ∼199°C (decomposition)
pH 6
Flash Point 270°C (518°F)
Assay Percent Range 98%
Odor Characteristic
Linear Formula ClCH2CO2Na
Quantity 10,000 g
UN Number UN2659
Beilstein 3597157
Merck Index 14,2112
Solubility Information Soluble in water,ether,chloroform,benzene and alcohol.
Formula Weight 116.48
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium chloroacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium chloroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium chloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.