missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice

Product Code. 15682980
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
6 x 250 g
50 kg
6 x 1 kg
Unit Size:
1kg
250g
500g
50kg
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15682980 50 kg 50kg
15662980 250 g 250g
15672980 500 g 500g
15652980 1 kg 1kg
15602990 6 x 250 g Pack of 6
15692980 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15682980 Supplier Honeywell-Fluka Supplier No. 3010550KG
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Boiling Point 1,704°C
Melting Point 993°C (literature)
CAS Max % 100.0000%
Quantity 50 kg
Linear Formula HCONH2
UN Number UN1690
IUPAC Name sodium fluoride
Formula Weight 41.99
Percent Purity >99%
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Packaging Fiberboard Box
Vapor Pressure 1.4mmHg (0°C)
Density 2.7 g/cm3
Chemical Name or Material Sodium fluoride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.