missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexafluorophosphate, TRC

Product Code. 30427396
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427396 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427396 Supplier Toronto Research Chemicals Supplier No. TRCS63431010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21324-39-0
Molecular Formula F6 P . Na
Molecular Weight (g/mol) 167.954
Synonym Sodium Hexafluorophosphate, Phosphorus Sodium Fluoride (PNaF6), Sodium Hexafluorophosphate (NaPF6), Sodium hexafluorophosphate(1-)
IUPAC Name sodium;hexafluorophosphate
SMILES [Na+].F[P-](F)(F)(F)(F)F

Specifications

Quantity 10 g
InChI Formula InChI=1S/F6P.Na/c1-7(2,3,4,5)6;/q-1;+1
Formula Weight 167.954
Chemical Name or Material Sodium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.