missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.0200 N (N/50), 1 mL = 1 mg CaCO3, Ricca Chemical

Product Code. 31750227
Change view
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
Unit Size:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31750227 20 L Cubitainer 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750227 Supplier Ricca Chemical Company Supplier No. R730000020F
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 98%, 0.16
CAS Max % 100%, 0.16
Concentration 0.01995 to 0.02005 N
Synonym sodium hydroxide
Quantity 20 L
IUPAC Name sodium hydroxide
Grade Laboratory
Packaging Cubitainer
Assay Normal at 20°C
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.