Learn More
Sodium Hydroxide, 0.1m, Reagecon™
Product Code. 11562982 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11562982 1 L 1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11562982 Supplier Reagecon Supplier No. S20101