missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 2.50 N, Ricca Chemical

Product Code. 31749908
Change view
Click to view available options
Quantity:
25 mL
10 L
Packaging:
25 mL HDPE Bottle
Cubitainer
Unit Size:
10 L
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31749908 25 mL 25 mL HDPE Bottle 25mL
31750468 10 L Cubitainer 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31749908 Supplier Ricca Chemical Company Supplier No. R745700025A
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Color Colorless
Boiling Point Approximately 100°C
Melting Point Approximately 0°C
CAS Max % 9.27%, 90.73%
CAS Min % 90.73%, 9.27%
Concentration 2.50 N
Concentration or Composition Notes 2.50 Normal
Quantity 25 mL
Grade Laboratory
Packaging 25 mL HDPE Bottle
Traceability to NIST NIST SRM# 723
Physical Form Liquid
Assay Normal at 20°C
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.