missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 2.50 N, Ricca Chemical

Product Code. 31750468
Change view
Click to view available options
Quantity:
25 mL
10 L
Packaging:
25 mL HDPE Bottle
Cubitainer
Unit Size:
10 L
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31750468 10 L Cubitainer 10 L
31749908 25 mL 25 mL HDPE Bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31750468 Supplier Ricca Chemical Company Supplier No. R745700010F
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
Concentration 2.499 to 2.501 N
Synonym sodium hydroxide
Quantity 10 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Grade Laboratory
Packaging Cubitainer
Physical Form Liquid
Assay Normal at 20°C
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.